Learn More
Genscript Corporation M-MuLV Reverse Transcriptase
Catalog No. 5000057093
Encompass
GenScript M-MuLV Reverse Transcriptase (M-MLV) is derived from a cloned region of the pol gene of MMLV and isolated from an E coli strain overexpressing this construct To increase cDNA yields and get a higher percentage of longer transcripts the M-MLV Reverse Transcriptase has been modified with reduced RNase H activity and expressed free of exogenous RNases and other nucleases The enzyme can synthesize a complementary cDNA strand initiating from a primer using RNA as template (cDNA synthesis) making it ideal for a wide range of applications
Catalog No. 50-000-57093 Supplier Genscript Corporation Supplier No. E0005010000
GenScript M-MuLV Reverse Transcriptase (M-MLV) is derived from a cloned region of the pol gene of MMLV and isolated from an E coli strain overexpressing this construct To increase cDNA yields and get a higher percentage of longer transcripts the M-MLV Reverse Transcriptase has been modified with reduced RNase H activity and expressed free of exogenous RNases and other nucleases The enzyme can synthesize a complementary cDNA strand initiating from a primer using RNA as template (cDNA synthesis) making it ideal for a wide range of applications