Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG HRP polymer Immunodetection Kit
SDP

Catalog No. MP2400NB
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP2400NB 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP2400NB Supplier Novus Biologicals™ Supplier No. MP2400NB

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Immunodetection Kit

Specifications

Quantity 1 Kit
Target Molecule M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG
Content And Storage Store at 4°C. Do not freeze.
For Use With (Application) Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.