Learn More
Medchemexpress LLC m-PEG-azide | 98.0% | 500 MG
SDP
Catalog No. 5000355470
Encompass_Preferred
m-PEG-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with Azide group
  • Enables CuAAc reactions with Alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • Utilizes ubiquitin-proteasome system for selective protein degradation
  • White to off-white solid
  • For research use only
Catalog No. 50-003-55470 Supplier Medchemexpress LLC Supplier No. HY140683500MG
m-PEG-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with Azide group
  • Enables CuAAc reactions with Alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • Utilizes ubiquitin-proteasome system for selective protein degradation
  • White to off-white solid
  • For research use only