Learn More
Medchemexpress LLC M-PEG-azide (MW 5000) | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1406831G
Encompass_Preferred
M-PEG-azide (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-55645