Learn More
Medchemexpress LLC m-PEG-azide (MW 5000) | 98.0% | COCCOCCN=[N+]=[N-].[n].[MW 5000] | 50 MG
SDP
Catalog No. 5000353774
Encompass_Preferred
m-PEG-azide (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker.
  • Click chemistry reagent.
  • Contains an Azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc).
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
Catalog No. 50-003-53774 Supplier Medchemexpress LLC Supplier No. HY14068350MG
m-PEG-azide (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker.
  • Click chemistry reagent.
  • Contains an Azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc).
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.