Learn More
Medchemexpress LLC M-PEG-C1-DSPE sodium | 1000 | 1 G
SDP
Catalog No. 5000365496
Encompass_Preferred
m-PEG-C1-DSPE sodium is a structural analog of m-PEG-DSPE sodium (HY-W440922). It is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. This product can be used in drug delivery research.
  • Improve the delivery efficiency of poorly soluble drugs
  • Improve the tissue specificity of poorly soluble drugs
  • Suitable for drug delivery research
Catalog No. 50-003-65496 Supplier Medchemexpress LLC Supplier No. 1GHYW440922A
m-PEG-C1-DSPE sodium is a structural analog of m-PEG-DSPE sodium (HY-W440922). It is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. This product can be used in drug delivery research.
  • Improve the delivery efficiency of poorly soluble drugs
  • Improve the tissue specificity of poorly soluble drugs
  • Suitable for drug delivery research