Learn More
Medchemexpress LLC M-PEG-C1-DSPE sodium | 1000 | 100 MG
SDP
Catalog No. 5000364744
Encompass_Preferred
m-PEG-C1-DSPE sodium (MW 1000) is a PEG lipid, serving as a structural analog to m-PEG-DSPE sodium (MW 1000). This compound is valuable in drug delivery research, particularly for improving the efficiency and tissue-specific delivery of drugs that are poorly soluble.
  • Enhances delivery efficiency of poorly soluble drugs.
  • Improves tissue specificity for drug targeting.
  • Suitable for various drug delivery research applications.
Catalog No. 50-003-64744 Supplier Medchemexpress LLC Supplier No. 100MGHYW440922A
m-PEG-C1-DSPE sodium (MW 1000) is a PEG lipid, serving as a structural analog to m-PEG-DSPE sodium (MW 1000). This compound is valuable in drug delivery research, particularly for improving the efficiency and tissue-specific delivery of drugs that are poorly soluble.
  • Enhances delivery efficiency of poorly soluble drugs.
  • Improves tissue specificity for drug targeting.
  • Suitable for various drug delivery research applications.