Learn More
Medchemexpress LLC M-PEG-C1-DSPE sodium (MW 1000) | 1000 | 250 MG
SDP
Catalog No. 5000366171
Encompass_Preferred
m-PEG-C1-DSPE sodium (MW 1000) is a structural analog of m-PEG-DSPE sodium (MW 1000) that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. This PEG lipid can be used in drug delivery research. It appears as a solid.
  • Structural analog of m-PEG-DSPE sodium (MW 1000)
  • Improves delivery efficiency and tissue specificity of poorly soluble drugs
  • Used in drug delivery research
Catalog No. 50-003-66171 Supplier Medchemexpress LLC Supplier No. 250MGHYW440922A
m-PEG-C1-DSPE sodium (MW 1000) is a structural analog of m-PEG-DSPE sodium (MW 1000) that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. This PEG lipid can be used in drug delivery research. It appears as a solid.
  • Structural analog of m-PEG-DSPE sodium (MW 1000)
  • Improves delivery efficiency and tissue specificity of poorly soluble drugs
  • Used in drug delivery research