Learn More
Medchemexpress LLC M-PEG-C1-DSPE sodium (MW 1000) | 1000 | 500 MG
SDP
Catalog No. 5000365910
Encompass_Preferred
M-PEG-C1-DSPE sodium (MW 1000) is a PEG lipid and a structural analog of m-PEG-DSPE sodium (MW 1000) (HY-W440922). It can improve the delivery efficiency and tissue specificity of poorly soluble drugs.
  • Enhances delivery efficiency of poorly soluble drugs.
  • Enhances tissue specificity of poorly soluble drugs.
  • Suitable for use in drug delivery research.
Catalog No. 50-003-65910 Supplier Medchemexpress LLC Supplier No. 500MGHYW440922A
M-PEG-C1-DSPE sodium (MW 1000) is a PEG lipid and a structural analog of m-PEG-DSPE sodium (MW 1000) (HY-W440922). It can improve the delivery efficiency and tissue specificity of poorly soluble drugs.
  • Enhances delivery efficiency of poorly soluble drugs.
  • Enhances tissue specificity of poorly soluble drugs.
  • Suitable for use in drug delivery research.