Learn More
Medchemexpress LLC M-PEG-DMG | 1019000-64-6 | 98.0% | 2000 Da (average) | 25 MG
SDP
Catalog No. 5000281190
Encompass_Preferred
m-PEG-DMG (MW 2000) is a methoxy-polyethylene glycol-dimyristoyl glycerol PEG-lipid used to formulate PEGylated liposomes and lipid nanoparticles for research. It combines a hydrophobic dimyristoyl glycerol anchor with an average 2000 Da PEG chain to stabilize lipid bilayers, improve colloidal stability, and extend particle circulation in aqueous formulations.
  • Provides a hydrophilic PEG spacer to sterically stabilize liposomes and nanoparticles.
  • Average molecular weight 2000 Da for predictable PEG coverage.
  • Dimyristoyl glycerol anchor promotes incorporation into lipid bilayers.
  • Improves colloidal stability and reduces particle aggregation.
  • Suitable for research use in drug delivery and formulation studies.
Catalog No. 50-002-81190 Supplier Medchemexpress LLC Supplier No. HYW44082125MG
m-PEG-DMG (MW 2000) is a methoxy-polyethylene glycol-dimyristoyl glycerol PEG-lipid used to formulate PEGylated liposomes and lipid nanoparticles for research. It combines a hydrophobic dimyristoyl glycerol anchor with an average 2000 Da PEG chain to stabilize lipid bilayers, improve colloidal stability, and extend particle circulation in aqueous formulations.
  • Provides a hydrophilic PEG spacer to sterically stabilize liposomes and nanoparticles.
  • Average molecular weight 2000 Da for predictable PEG coverage.
  • Dimyristoyl glycerol anchor promotes incorporation into lipid bilayers.
  • Improves colloidal stability and reduces particle aggregation.
  • Suitable for research use in drug delivery and formulation studies.