Learn More
Medchemexpress LLC m-PEG-DMG | 1019000-64-6 | ≥98.0% | 2000 Da (average) | (C2H4O)nC36H73NO5 | 5 MG
SDP
Catalog No. 5000281167
Encompass_Preferred
m-PEG-DMG (MW 2000) is a methoxy-terminated polyethylene glycol lipid with a dimyristoyl glycerol anchor and an average molecular weight of 2,000 Da. It is intended for incorporation into liposomes and lipid nanoparticles to provide a PEG corona that enhances stability and circulation time in drug delivery research.
  • Enhances colloidal stability and circulation time
  • Reduces protein adsorption and opsonization
  • Facilitates PEGylation of liposome and nanoparticle surfaces
  • Supplied in lab-scale quantities suitable for formulation studies
  • High purity for reproducible formulation performance
Catalog No. 50-002-81167 Supplier Medchemexpress LLC Supplier No. HYW4408215MG
m-PEG-DMG (MW 2000) is a methoxy-terminated polyethylene glycol lipid with a dimyristoyl glycerol anchor and an average molecular weight of 2,000 Da. It is intended for incorporation into liposomes and lipid nanoparticles to provide a PEG corona that enhances stability and circulation time in drug delivery research.
  • Enhances colloidal stability and circulation time
  • Reduces protein adsorption and opsonization
  • Facilitates PEGylation of liposome and nanoparticle surfaces
  • Supplied in lab-scale quantities suitable for formulation studies
  • High purity for reproducible formulation performance