Learn More
Medchemexpress LLC m-PEG-DMG (MW 2000) | 1019000-64-6 | 98.0% | 2000 (average) | No fixed chemical formula - polymeric m-PEG-DMG (average MW 2000) | 10 MG
SDP
Catalog No. 5000280537
Encompass_Preferred
m-PEG-DMG (MW 2000) is a methoxy-terminated polyethylene glycol conjugated to dimyristoyl glycerol, provided as a solid PEGylated lipid for formulation research. It is used to prepare liposomes and lipid nanoparticles for drug delivery studies, and is supplied at an average molecular weight of 2000 with high purity for reliable experimental results.
  • PEGylated lipid for liposome and lipid nanoparticle formulation.
  • Average molecular weight 2000 for consistent PEG chain length.
  • High purity (~98.0%) suitable for research applications.
  • White to off-white solid, easy to handle in laboratory settings.
  • Stable when stored as a powder at low temperatures for extended shelf life.
Catalog No. 50-002-80537 Supplier Medchemexpress LLC Supplier No. HYW44082110MG
m-PEG-DMG (MW 2000) is a methoxy-terminated polyethylene glycol conjugated to dimyristoyl glycerol, provided as a solid PEGylated lipid for formulation research. It is used to prepare liposomes and lipid nanoparticles for drug delivery studies, and is supplied at an average molecular weight of 2000 with high purity for reliable experimental results.
  • PEGylated lipid for liposome and lipid nanoparticle formulation.
  • Average molecular weight 2000 for consistent PEG chain length.
  • High purity (~98.0%) suitable for research applications.
  • White to off-white solid, easy to handle in laboratory settings.
  • Stable when stored as a powder at low temperatures for extended shelf life.