Learn More
Medchemexpress LLC M-PEG-DSPE sodium | 95.0% | 3000 (average) | 25 MG
SDP
Catalog No. 5000281473
Encompass_Preferred
m-PEG-DSPE sodium (MW 3000) is a methoxy polyethylene glycol-distearoylphosphatidylethanolamine (PEG-lipid) provided as the sodium salt for use in drug delivery research. It is formulated to act as a surface-modifying lipid that improves colloidal stability and tissue targeting when incorporated into liposomes or nanoparticles.
  • Provides hydrophilic PEG coating to reduce protein adsorption and extend circulation time.
  • Average molecular weight of 3000 for predictable PEG chain length.
  • Solid, white to off-white material suitable for formulation.
  • High purity (95.0%) for reproducible experimental results.
  • Compatible with nanoparticle and liposome formulation workflows.
  • Store sealed at -20°C; solvent storage guidance for longer-term stability.
Catalog No. 50-002-81473 Supplier Medchemexpress LLC Supplier No. HYW44092425MG
m-PEG-DSPE sodium (MW 3000) is a methoxy polyethylene glycol-distearoylphosphatidylethanolamine (PEG-lipid) provided as the sodium salt for use in drug delivery research. It is formulated to act as a surface-modifying lipid that improves colloidal stability and tissue targeting when incorporated into liposomes or nanoparticles.
  • Provides hydrophilic PEG coating to reduce protein adsorption and extend circulation time.
  • Average molecular weight of 3000 for predictable PEG chain length.
  • Solid, white to off-white material suitable for formulation.
  • High purity (95.0%) for reproducible experimental results.
  • Compatible with nanoparticle and liposome formulation workflows.
  • Store sealed at -20°C; solvent storage guidance for longer-term stability.