Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG-DSPE (sodium) | 95.0% | 3000 (average) | C45H88NO11P | 1 G
SDP

Catalog No. 5000281193
Encompass_Preferred

m-PEG-DSPE sodium (MW 3000) is a PEGylated phospholipid supplied as a white to off-white solid. The sodium salt of methoxy polyethylene glycol-distearoylphosphoethanolamine (average PEG MW 3,000 Da), it is used to stabilize liposomes and lipid nanoparticles, provide steric stabilization, and modify particle surfaces for drug delivery research.

  • Average molecular weight 3000 Da.
  • Purity 95.0%.
  • White to off-white solid form.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Provides steric stabilization and surface PEGylation.
  • Storage: -20°C sealed; in solvent: -80°C up to 6 months.
  • Available in laboratory pack sizes including 1 g.

Catalog No. 50-002-81193 Supplier Medchemexpress LLC Supplier No. HYW4409241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG-DSPE sodium (MW 3000) is a PEGylated phospholipid supplied as a white to off-white solid. The sodium salt of methoxy polyethylene glycol-distearoylphosphoethanolamine (average PEG MW 3,000 Da), it is used to stabilize liposomes and lipid nanoparticles, provide steric stabilization, and modify particle surfaces for drug delivery research.

  • Average molecular weight 3000 Da.
  • Purity 95.0%.
  • White to off-white solid form.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Provides steric stabilization and surface PEGylation.
  • Storage: -20°C sealed; in solvent: -80°C up to 6 months.
  • Available in laboratory pack sizes including 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.