Learn More
Medchemexpress LLC M-PEG-mal (MW 2000) | 97.27% | 100 MG
SDP
Catalog No. 5000356474
Encompass_Preferred
M-PEG-mal (MW 2000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are innovative molecules that harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins, utilizing two different ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for the protein of interest.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56474 Supplier Medchemexpress LLC Supplier No. HY140689100MG
M-PEG-mal (MW 2000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are innovative molecules that harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins, utilizing two different ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for the protein of interest.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins