Learn More
Medchemexpress LLC m-PEG-mal (MW 30000) | 98.0% | 100 MG
SDP
Catalog No. 5000355469
Encompass_Preferred
m-PEG-mal (MW 30000) is a PEG-based PROTAC linker for the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and not for patient use.
  • PEG-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Utilizes the ubiquitin-proteasome system to degrade target proteins
  • Related to cancer and cancer targeted therapy
Catalog No. 50-003-55469 Supplier Medchemexpress LLC Supplier No. HY140693100MG
m-PEG-mal (MW 30000) is a PEG-based PROTAC linker for the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and not for patient use.
  • PEG-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Utilizes the ubiquitin-proteasome system to degrade target proteins
  • Related to cancer and cancer targeted therapy