Learn More
Medchemexpress LLC m-PEG-mal (MW 30000) | 98.0% | 250 MG
SDP
Catalog No. 5000352586
Encompass_Preferred
m-PEG-mal (MW 30000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only in laboratory settings. PROTACs are designed to utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • For synthesis of PROTACs
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52586 Supplier Medchemexpress LLC Supplier No. 250MGHY140693
m-PEG-mal (MW 30000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only in laboratory settings. PROTACs are designed to utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • For synthesis of PROTACs
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only