Learn More
Medchemexpress LLC M-PEG-mal (MW 30000) | 98.0% | 500 MG
SDP
Catalog No. 5000352931
Encompass_Preferred
M-PEG-mal (MW 30000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Degrades target proteins selectively
  • Features two ligands connected by a linker
  • One ligand for E3 ubiquitin ligase, one for target protein
Catalog No. 50-003-52931 Supplier Medchemexpress LLC Supplier No. HY140693500MG
M-PEG-mal (MW 30000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Degrades target proteins selectively
  • Features two ligands connected by a linker
  • One ligand for E3 ubiquitin ligase, one for target protein