Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG-NH2 hydrochloride | 99.1% | 2000 (Mn) | 50 MG
SDP

Catalog No. 5000284003
Encompass_Preferred

m-PEG-NH2 hydrochloride (Mn 2000) is a methoxy-terminated polyethylene glycol bearing a terminal primary amine as its hydrochloride salt. Supplied as a solid reagent, it is used for PEGylation, surface functionalization, and preparation of gels for adsorption-size-exclusion (AdSEC) separations in biochemical workflows.

  • Primary amine hydrochloride form for conjugation reactions.
  • Average molecular weight Mn 2000 for consistent polymer properties.
  • High purity: 99.13%.
  • Soluble in DMSO at 100 mg/mL with sonication (use dry DMSO).
  • Storage: store at -20°C, protect from light; in solution store at -80°C up to 6 months.
  • For research use only.

Catalog No. 50-002-84003 Supplier Medchemexpress LLC Supplier No. HYW598230A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG-NH2 hydrochloride (Mn 2000) is a methoxy-terminated polyethylene glycol bearing a terminal primary amine as its hydrochloride salt. Supplied as a solid reagent, it is used for PEGylation, surface functionalization, and preparation of gels for adsorption-size-exclusion (AdSEC) separations in biochemical workflows.

  • Primary amine hydrochloride form for conjugation reactions.
  • Average molecular weight Mn 2000 for consistent polymer properties.
  • High purity: 99.13%.
  • Soluble in DMSO at 100 mg/mL with sonication (use dry DMSO).
  • Storage: store at -20°C, protect from light; in solution store at -80°C up to 6 months.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.