Learn More
Medchemexpress LLC m-PEG-NH2 hydrochloride (MW 2000) | 99.1% | 2000 (Average) | 250 MG
SDP
Catalog No. 5000284259
Encompass_Preferred
m-PEG-NH2 hydrochloride (MW 2000) is a methoxy-terminated polyethylene glycol bearing a terminal primary amine, supplied as the hydrochloride salt. It is used as a reactive PEG building block for PEGylation, bioconjugation, hydrogel synthesis, and formulation of adsorption/size-exclusion gels for biological separations.
  • Average molecular weight 2000 (average).
  • Terminal primary amine as hydrochloride salt.
  • High purity (~99.1%).
  • Suitable for PEGylation and bioconjugation reactions.
  • Compatible with hydrogel and AdSEC gel formulation.
  • Available in research sizes such as 250 MG.
Catalog No. 50-002-84259 Supplier Medchemexpress LLC Supplier No. HYW598230A250MG
m-PEG-NH2 hydrochloride (MW 2000) is a methoxy-terminated polyethylene glycol bearing a terminal primary amine, supplied as the hydrochloride salt. It is used as a reactive PEG building block for PEGylation, bioconjugation, hydrogel synthesis, and formulation of adsorption/size-exclusion gels for biological separations.
  • Average molecular weight 2000 (average).
  • Terminal primary amine as hydrochloride salt.
  • High purity (~99.1%).
  • Suitable for PEGylation and bioconjugation reactions.
  • Compatible with hydrogel and AdSEC gel formulation.
  • Available in research sizes such as 250 MG.