Learn More
Medchemexpress LLC M-PEG-NHS ester | 92451-01-9 | 98.0% | (C2H4O)nC7H9NO5 | 50 MG
SDP
Catalog No. 5000353130
Encompass_Preferred
M-PEG-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Protacs contain two different ligands connected by a linker
  • Features a ligand for an E3 ubiquitin ligase and another for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53130 Supplier Medchemexpress LLC Supplier No. HY14070050MG
M-PEG-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Protacs contain two different ligands connected by a linker
  • Features a ligand for an E3 ubiquitin ligase and another for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins