Learn More
Medchemexpress LLC M-PEG-NHS ester | 92451-01-9 | 98.0% | Poly(oxy-1,2-ethanediyl), α-[2-[(2,5-dioxo-1-pyrrolidinyl)oxy]-2-oxoethyl]-ω-methoxy- | 100 MG
SDP
Catalog No. 5000352936
Encompass_Preferred
M-PEG-NHS ester (MW 10000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52936 Supplier Medchemexpress LLC Supplier No. HY140699100MG
M-PEG-NHS ester (MW 10000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins