Learn More
Medchemexpress LLC m-PEG-NHS ester | 92451-01-9 | 98.0% | Poly(oxy-1,2-ethanediyl), α-[2-[(2,5-dioxo-1-pyrrolidinyl)oxy]-2-oxoethyl]-ω-methoxy- | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140699
Encompass_Preferred
m-PEG-NHS ester (MW 10000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein, thereby leveraging the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Targets PROTAC linkers
  • Exploits the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-57251