Learn More
Medchemexpress LLC M-PEG-NHS ester | 92451-01-9 | MFCD01867746 | 98.0% | 2000 (average) | (C2H4O)nC7H9NO5 | 100 MG
SDP
Catalog No. 5000285261
Encompass_Preferred
m-PEG-NHS ester (MW 2000) is an NHS-activated methoxy poly(ethylene glycol) reagent for PEGylation of biomolecules. It reacts with primary amines to modify proteins, peptides, and small molecules to improve solubility, stability, and pharmacokinetics. Supplied as a white to off-white solid for research use, it is offered in multiple pack sizes and stored sealed away from moisture.
Catalog No. 50-002-85261 Supplier Medchemexpress LLC Supplier No. HYW591424100MG
m-PEG-NHS ester (MW 2000) is an NHS-activated methoxy poly(ethylene glycol) reagent for PEGylation of biomolecules. It reacts with primary amines to modify proteins, peptides, and small molecules to improve solubility, stability, and pharmacokinetics. Supplied as a white to off-white solid for research use, it is offered in multiple pack sizes and stored sealed away from moisture.