Learn More
Medchemexpress LLC m-PEG-NHS ester | 92451-01-9 | MFCD06412227 | 98.0% | 2,000 Da | C9H13NO6 | 500 MG
SDP
Catalog No. 5000284401
Encompass_Preferred
m-PEG-NHS ester is a methoxy-terminated polyethylene glycol reagent functionalized with an N-hydroxysuccinimide (NHS) ester for selective conjugation to primary amines. With an average molecular weight of approximately 2,000 Da, it is used to introduce PEG chains that improve solubility, stability, and circulation time of proteins, peptides, and small molecules. Supplied as a 500 mg solid.
  • Methoxy-terminated PEG with NHS ester reactive group.
  • Average molecular weight ~2,000 Da for spacer and solubility enhancement.
  • High purity suitable for bioconjugation (reported 98.0%).
  • Reacts selectively with primary amines to form stable amide linkages.
  • Commonly used for PEGylation, linker construction, and bioconjugation.
Catalog No. 50-002-84401 Supplier Medchemexpress LLC Supplier No. HYW591424500MG
m-PEG-NHS ester is a methoxy-terminated polyethylene glycol reagent functionalized with an N-hydroxysuccinimide (NHS) ester for selective conjugation to primary amines. With an average molecular weight of approximately 2,000 Da, it is used to introduce PEG chains that improve solubility, stability, and circulation time of proteins, peptides, and small molecules. Supplied as a 500 mg solid.
  • Methoxy-terminated PEG with NHS ester reactive group.
  • Average molecular weight ~2,000 Da for spacer and solubility enhancement.
  • High purity suitable for bioconjugation (reported 98.0%).
  • Reacts selectively with primary amines to form stable amide linkages.
  • Commonly used for PEGylation, linker construction, and bioconjugation.