Learn More
Medchemexpress LLC m-PEG-NHS ester (MW 10000) | 92451-01-9 | 98.0% | Poly(oxy-1,2-ethanediyl), α-[2-[(2,5-dioxo-1-pyrrolidinyl)oxy]-2-oxoethyl]-ω-methoxy- | 500 MG
SDP
Catalog No. 5000356632
Encompass_Preferred
m-PEG-NHS ester (MW 10000) is a PEG-based PROTAC linker that facilitates the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. This product is intended for research purposes only.
  • PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
  • Suitable for research use only
Catalog No. 50-003-56632 Supplier Medchemexpress LLC Supplier No. HY140699500MG
m-PEG-NHS ester (MW 10000) is a PEG-based PROTAC linker that facilitates the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. This product is intended for research purposes only.
  • PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
  • Suitable for research use only