Learn More
Medchemexpress LLC M-PEG-NHS ester (MW 5000) | 92451-01-9 | 95.0% | (C2H4O)nC7H9NO5 | 100 MG
SDP
Catalog No. 5000355135
Encompass_Preferred
m-PEG-NHS ester (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system. It has an average molecular weight of 5000 and appears as a white to off-white solid. This product is for research use only.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Average molecular weight of 5000
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-003-55135 Supplier Medchemexpress LLC Supplier No. HY140698100MG
m-PEG-NHS ester (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system. It has an average molecular weight of 5000 and appears as a white to off-white solid. This product is for research use only.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Average molecular weight of 5000
  • White to off-white solid appearance
  • For research use only