Learn More
Medchemexpress LLC m-PEG-NHS ester (MW 5000) | 92451-01-9 | 95.0% | (C2H4O)nC7H9NO5 | 50 MG
SDP
Catalog No. 5000352935
Encompass_Preferred
m-PEG-NHS ester (MW 5000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This solid, white to off-white compound boasts a purity of 95.0% and is intended strictly for research purposes.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Solid appearance
  • White to off-white color
  • 95.0% purity
Catalog No. 50-003-52935 Supplier Medchemexpress LLC Supplier No. HY14069850MG
m-PEG-NHS ester (MW 5000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This solid, white to off-white compound boasts a purity of 95.0% and is intended strictly for research purposes.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Solid appearance
  • White to off-white color
  • 95.0% purity