Learn More
Medchemexpress LLC M-PEG-thiol | 134874-49-0 | MFCD21363241 | 98.7% | (C2H4O)nC3H8OS | 50 MG
SDP
Catalog No. 5000284487
Encompass_Preferred
m-PEG-thiol (MW 1000) is a methoxy-terminated polyethylene glycol bearing a terminal thiol group. Supplied as a white to light yellow solid with an average molecular weight of approximately 1000 Da, it is used to introduce thiol functionality for surface modification, nanoparticle functionalization, and bioconjugation.
  • Provides a terminal thiol group for gold-thiol conjugation and thiol-maleimide coupling.
  • Suitable for surface modification of nanoparticles and thiolation of nucleic acids.
  • High purity (98.66%) supporting reproducible conjugation chemistry.
  • Physical form solid; color white to light yellow.
  • Available in 50 MG and larger pack sizes.
Catalog No. 50-002-84487 Supplier Medchemexpress LLC Supplier No. HYW59147650MG
m-PEG-thiol (MW 1000) is a methoxy-terminated polyethylene glycol bearing a terminal thiol group. Supplied as a white to light yellow solid with an average molecular weight of approximately 1000 Da, it is used to introduce thiol functionality for surface modification, nanoparticle functionalization, and bioconjugation.
  • Provides a terminal thiol group for gold-thiol conjugation and thiol-maleimide coupling.
  • Suitable for surface modification of nanoparticles and thiolation of nucleic acids.
  • High purity (98.66%) supporting reproducible conjugation chemistry.
  • Physical form solid; color white to light yellow.
  • Available in 50 MG and larger pack sizes.