Learn More
Medchemexpress LLC m-PEG-thiol (MW 1000) | 134874-49-0 | 98.0% | (C2H4O)nC3H8OS | 25 MG
SDP
Catalog No. 5000357093
Encompass_Preferred
m-PEG-thiol (MW 1000) is a surface modifier used to modify DNA thiolation and in the synthesis of gold nanorods (AuNR). It forms a covalent bond with the surface of gold nanoparticles through its thiol group, stabilizing them and enhancing biocompatibility. This substance provides a platform for subsequent coupling of biomolecules, promoting the functionalization of nanomaterials.
  • Used for surface functionalization of gold nanorods (AuNRs)
  • Modifies DNA thiolation
  • Stabilizes nanoparticles by steric hindrance effect
  • Enhances biocompatibility of nanoparticles
  • Provides a platform for coupling biomolecules (such as DNA, antibodies)
  • Can be used as a shielding layer to embed Raman reporter molecules or drugs
  • Suitable for SERS imaging of cancer cells and construction of drug delivery systems
Catalog No. 50-003-57093 Supplier Medchemexpress LLC Supplier No. 25MGHY140708
m-PEG-thiol (MW 1000) is a surface modifier used to modify DNA thiolation and in the synthesis of gold nanorods (AuNR). It forms a covalent bond with the surface of gold nanoparticles through its thiol group, stabilizing them and enhancing biocompatibility. This substance provides a platform for subsequent coupling of biomolecules, promoting the functionalization of nanomaterials.
  • Used for surface functionalization of gold nanorods (AuNRs)
  • Modifies DNA thiolation
  • Stabilizes nanoparticles by steric hindrance effect
  • Enhances biocompatibility of nanoparticles
  • Provides a platform for coupling biomolecules (such as DNA, antibodies)
  • Can be used as a shielding layer to embed Raman reporter molecules or drugs
  • Suitable for SERS imaging of cancer cells and construction of drug delivery systems