Learn More
Medchemexpress LLC m-PEG-thiol (MW 10000) | 98.0% | 100 MG
SDP
Catalog No. 5000354941
Encompass_Preferred
m-PEG-thiol (MW 10000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
Catalog No. 50-003-54941 Supplier Medchemexpress LLC Supplier No. HY140708100MG
m-PEG-thiol (MW 10000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere