Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG-thiol (MW 10000) | 98.0% | 50 MG
SDP

Catalog No. 5000353790
Encompass_Preferred

m-PEG-thiol (MW 10000), also known as mPEG-SH (MW 10000), is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the ubiquitin-proteasome system for targeted protein degradation.

  • Purity: 98.0%
  • Molecular weight: 10000 (average)
  • Appearance: Solid, white to off-white
  • Application: Used in the synthesis of PROTACs for targeted protein degradation research
  • Storage: 4°C, stored under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month (under nitrogen)

Catalog No. 50-003-53790 Supplier Medchemexpress LLC Supplier No. HY14070850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG-thiol (MW 10000), also known as mPEG-SH (MW 10000), is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the ubiquitin-proteasome system for targeted protein degradation.

  • Purity: 98.0%
  • Molecular weight: 10000 (average)
  • Appearance: Solid, white to off-white
  • Application: Used in the synthesis of PROTACs for targeted protein degradation research
  • Storage: 4°C, stored under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month (under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.