Learn More
Medchemexpress LLC M-PEG-thiol (MW 2000) | 134874-49-0 | 99.8% | 100 MG
SDP
Catalog No. 5000352934
Encompass_Preferred
M-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product has an average molecular weight of 2000 and a high purity of 99.8%.
  • Appears as a white to yellow solid
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Can be shipped at room temperature in the continental US
Catalog No. 50-003-52934 Supplier Medchemexpress LLC Supplier No. HY140706100MG
M-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product has an average molecular weight of 2000 and a high purity of 99.8%.
  • Appears as a white to yellow solid
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Can be shipped at room temperature in the continental US