Learn More
Medchemexpress LLC m-PEG-thiol (MW 2000) | 134874-49-0 | 99.8% | 250 MG
SDP
Catalog No. 5000352756
Encompass_Preferred
m-PEG-thiol (MW 2000) is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrade target proteins
Catalog No. 50-003-52756 Supplier Medchemexpress LLC Supplier No. 250MGHY140709
m-PEG-thiol (MW 2000) is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrade target proteins