Learn More
Medchemexpress LLC M-PEG-thiol (MW 2000) | 134874-49-0 | 99.8% | COCCOCCS.[MW 2000].[n] | 50 MG
SDP
Catalog No. 5000356629
Encompass_Preferred
M-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules feature two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for the target protein. PROTACs harness the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56629 Supplier Medchemexpress LLC Supplier No. HY14070650MG
M-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules feature two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for the target protein. PROTACs harness the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins