Learn More
Medchemexpress LLC M-PEG-thiol (MW 2000) | 134874-49-0 | 99.8% | COCCOCCS.[MW 2000].[n] | 500 MG
SDP
Catalog No. 5000353156
Encompass_Preferred
M-PEG-thiol (MW 2000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-53156 Supplier Medchemexpress LLC Supplier No. HY140709500MG
Add to Cart
Add to Cart
M-PEG-thiol (MW 2000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only