Learn More
Medchemexpress LLC m-PEG-thiol (MW 20000) | 95.0% | 100 MG
SDP
Catalog No. 5000352961
Encompass_Preferred
m-PEG-thiol (MW 20000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed with two ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-52961 Supplier Medchemexpress LLC Supplier No. HY140709100MG
m-PEG-thiol (MW 20000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed with two ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins