Learn More
Medchemexpress LLC m-PEG-thiol (MW 20000) | 95.0% | 50 MG
SDP
Catalog No. 5000354939
Encompass_Preferred
m-PEG-thiol (MW 20000) is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs operate by connecting two different ligands, one for an E3 ubiquitin ligase and another for a target protein, to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is for research use only and has not been validated for medical applications.
  • PEG-based PROTAC linker
  • Designed for PROTAC synthesis
  • Facilitates selective degradation of target proteins
  • Average molecular weight of 20000
  • Appears as a white to off-white solid
  • Purity of 95.0%
Catalog No. 50-003-54939 Supplier Medchemexpress LLC Supplier No. HY14070950MG
m-PEG-thiol (MW 20000) is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs operate by connecting two different ligands, one for an E3 ubiquitin ligase and another for a target protein, to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is for research use only and has not been validated for medical applications.
  • PEG-based PROTAC linker
  • Designed for PROTAC synthesis
  • Facilitates selective degradation of target proteins
  • Average molecular weight of 20000
  • Appears as a white to off-white solid
  • Purity of 95.0%