Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG-thiol (MW 5000) | 88778-21-6 | 99.9% | 1 G
SDP

Catalog No. 5000356477
Encompass_Preferred

m-PEG-thiol (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This product functions by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Purity 99.9%
  • Molecular weight 5000 (Average)

Catalog No. 50-003-56477 Supplier Medchemexpress LLC Supplier No. HY1407071G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG-thiol (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This product functions by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Purity 99.9%
  • Molecular weight 5000 (Average)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.