Learn More
Medchemexpress LLC M-PEG-thiol (MW 5000) | 88778-21-6 | 99.9% | 500 MG
SDP
Catalog No. 5000353356
Encompass_Preferred
M-PEG-thiol (MW 5000) is a PEG-based PROTAC linker employed in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two different ligands: one for an E3 ubiquitin ligase and the other for the target protein.
  • Utilized as a PEG-based PROTAC linker
  • Supports the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-53356 Supplier Medchemexpress LLC Supplier No. HY140707500MG
M-PEG-thiol (MW 5000) is a PEG-based PROTAC linker employed in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two different ligands: one for an E3 ubiquitin ligase and the other for the target protein.
  • Utilized as a PEG-based PROTAC linker
  • Supports the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins