Learn More
Medchemexpress LLC m-PEG-triethoxysilane (MW 5000) | 547701-35-9 | 98.0% | C15H34N2O6Si | 250 MG
SDP
Catalog No. 5000352848
Encompass_Preferred
m-PEG-triethoxysilane (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules are designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. It has an average molecular weight of 5000 and appears as a white to off-white solid.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Appears as a white to off-white solid
  • Stable for up to 3 years as powder at -20°C
Catalog No. 50-003-52848 Supplier Medchemexpress LLC Supplier No. 250MGHY140704
m-PEG-triethoxysilane (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules are designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. It has an average molecular weight of 5000 and appears as a white to off-white solid.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Appears as a white to off-white solid
  • Stable for up to 3 years as powder at -20°C