Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG1-NHS ester 1g | 1027371-75-0 | 201.18 g/mol | 1 G
SDP

Catalog No. 5000230064
Encompass_Preferred

m-PEG1-NHS ester is a methoxy-terminated polyethylene glycol N-hydroxysuccinimide (NHS) ester used as a short PEG linker and an amine-reactive coupling reagent for bioconjugation and linker assembly in medicinal chemistry.

  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Short PEG spacer (approx. 201.18 g/mol) that improves solubility and reduces steric hindrance.
  • Supplied as a solid with recommended powder storage at -20°C to maintain stability.
  • Soluble in water and PBS with ultrasonic warming; H2O 33.33 mg/mL, PBS 100 mg/mL.
  • Purity reported as 95.0% for typical batches.

Catalog No. 50-002-30064 Supplier Medchemexpress LLC Supplier No. HY1145121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG1-NHS ester is a methoxy-terminated polyethylene glycol N-hydroxysuccinimide (NHS) ester used as a short PEG linker and an amine-reactive coupling reagent for bioconjugation and linker assembly in medicinal chemistry.

  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Short PEG spacer (approx. 201.18 g/mol) that improves solubility and reduces steric hindrance.
  • Supplied as a solid with recommended powder storage at -20°C to maintain stability.
  • Soluble in water and PBS with ultrasonic warming; H2O 33.33 mg/mL, PBS 100 mg/mL.
  • Purity reported as 95.0% for typical batches.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.