Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG1-NHS ester 250mg | 1027371-75-0 | 201.18 g/mol | C8H11NO5 | 250 MG
SDP

Supplier:  Medchemexpress LLC HY114512250MG

Encompass_Preferred

m-PEG1-NHS ester is a PEG/alkyl/ether-based N-hydroxysuccinimide (NHS) ester linker used for assembling proteolysis targeting chimeras (PROTACs) and for amine-directed bioconjugation. Supplied as a solid, it provides a short PEG spacer with an activated ester for efficient coupling in synthetic workflows.

  • PEG/alkyl/ether-based linker for PROTAC synthesis.
  • N-hydroxysuccinimide ester reactive toward primary amines.
  • Molecular weight 201.18 g/mol.
  • Chemical formula C8H11NO5.
  • Purity 95.0%.
  • Appearance white to light yellow solid.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available package size 250 MG.

Catalog No. 50-002-26693


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.