Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG10-acid | 2409969-94-2 | 98.0% | 500.58 | C22H44O12 | 25 MG
SDP

Catalog No. 5000203761
Encompass_Preferred

m-PEG10-acid is a non-cleavable polyethylene glycol (PEG) linker containing 10 ethylene glycol units. It is supplied as an oil and is used as a hydrophilic spacer in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs).

  • Non-cleavable 10-unit PEG structure suitable for linker applications.
  • Molecular weight 500.58 g·mol⁻¹ and formula C22H44O12.
  • Purity 98.0% by NMR.
  • Storage at -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, e.g., 25 mg.

Catalog No. 50-002-03761 Supplier Medchemexpress LLC Supplier No. HY14050025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG10-acid is a non-cleavable polyethylene glycol (PEG) linker containing 10 ethylene glycol units. It is supplied as an oil and is used as a hydrophilic spacer in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs).

  • Non-cleavable 10-unit PEG structure suitable for linker applications.
  • Molecular weight 500.58 g·mol⁻¹ and formula C22H44O12.
  • Purity 98.0% by NMR.
  • Storage at -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, e.g., 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.