Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG10-alcohol | 27425-92-9 | MFCD06797087 | >97.0% | 472.57 g/mol | C21H44O11 | 1 G
SDP

Catalog No. 5000203787
Encompass_Preferred

m-PEG10-alcohol (decaethylene glycol monomethyl ether) is a 10-unit monomethyl polyethylene glycol (PEG) reagent used as a non-cleavable PEG linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker for PROTAC construction. It is provided as a research reagent with characterized purity and defined storage recommendations.

  • Non-cleavable 10-unit PEG linker.
  • Synonym: decaethylene glycol monomethyl ether.
  • Purity ≥97.0%.
  • Molecular weight 472.57 g/mol, formula C21H44O11.
  • Available pack sizes: 500 mg, 1 g, 5 g, 10 g.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-03787 Supplier Medchemexpress LLC Supplier No. HY1412181G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG10-alcohol (decaethylene glycol monomethyl ether) is a 10-unit monomethyl polyethylene glycol (PEG) reagent used as a non-cleavable PEG linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker for PROTAC construction. It is provided as a research reagent with characterized purity and defined storage recommendations.

  • Non-cleavable 10-unit PEG linker.
  • Synonym: decaethylene glycol monomethyl ether.
  • Purity ≥97.0%.
  • Molecular weight 472.57 g/mol, formula C21H44O11.
  • Available pack sizes: 500 mg, 1 g, 5 g, 10 g.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.