Learn More
Medchemexpress LLC m-PEG11-acid | 2280998-74-3 | 98.0% | 544.63 | C24H48O13 | 100 MG
SDP
Catalog No. 5000203128
Encompass_Preferred
m-PEG11-acid is a monomethyl-capped polyethylene glycol (PEG) linker containing 11 ethylene glycol units. It is supplied as a non-cleavable, hydrophilic spacer for use in antibody-drug conjugate (ADC) synthesis and proteolysis-targeting chimera (PROTAC) linker construction.
  • Monomethyl-capped polyethylene glycol linker with 11 ethylene glycol units.
  • Non-cleavable design suitable for stable conjugation.
  • Provides a hydrophilic spacer to improve solubility and reduce steric hindrance.
  • Commonly used in ADC and PROTAC linker synthesis.
  • Molecular weight approximately 544.63 g/mol.
  • High purity, specified at 98.0%.
  • Available in small-scale packaging suitable for research (100 MG).
Catalog No. 50-002-03128 Supplier Medchemexpress LLC Supplier No. HY140501100MG
m-PEG11-acid is a monomethyl-capped polyethylene glycol (PEG) linker containing 11 ethylene glycol units. It is supplied as a non-cleavable, hydrophilic spacer for use in antibody-drug conjugate (ADC) synthesis and proteolysis-targeting chimera (PROTAC) linker construction.
  • Monomethyl-capped polyethylene glycol linker with 11 ethylene glycol units.
  • Non-cleavable design suitable for stable conjugation.
  • Provides a hydrophilic spacer to improve solubility and reduce steric hindrance.
  • Commonly used in ADC and PROTAC linker synthesis.
  • Molecular weight approximately 544.63 g/mol.
  • High purity, specified at 98.0%.
  • Available in small-scale packaging suitable for research (100 MG).