Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG11-acid | 2280998-74-3 | 98.0% | 544.63 g·mol⁻¹ | C24H48O13 | 50 MG
SDP

Catalog No. 5000201894
Encompass_Preferred

m-PEG11-acid is a monomethyl polyethylene glycol (11 ethylene glycol units) acid linker used as a reagent for antibody-drug conjugate and PROTAC linker synthesis, and for general bioconjugation.

  • Monomethyl PEG with 11 ethylene glycol units.
  • Molecular weight 544.63 g·mol⁻¹.
  • Purity 98.0%.
  • Cas number 2280998-74-3.
  • Available in small pack sizes, including 50 mg and 100 mg.

Catalog No. 50-002-01894 Supplier Medchemexpress LLC Supplier No. HY14050150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG11-acid is a monomethyl polyethylene glycol (11 ethylene glycol units) acid linker used as a reagent for antibody-drug conjugate and PROTAC linker synthesis, and for general bioconjugation.

  • Monomethyl PEG with 11 ethylene glycol units.
  • Molecular weight 544.63 g·mol⁻¹.
  • Purity 98.0%.
  • Cas number 2280998-74-3.
  • Available in small pack sizes, including 50 mg and 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.