Learn More
Medchemexpress LLC m-PEG11-NHS ester | 3119903-67-9 | C28H51NO15 | 100 MG
SDP
Catalog No. 5000355056
Encompass_Preferred
m-PEG11-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is recommended for storage at -20°C, sealed, and away from moisture and light. If in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also with sealed storage, away from moisture and light.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Store at -20°C, sealed, away from moisture and light
  • Extended storage in solvent possible at -80°C
Catalog No. 50-003-55056 Supplier Medchemexpress LLC Supplier No. HY141105100MG
m-PEG11-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is recommended for storage at -20°C, sealed, and away from moisture and light. If in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also with sealed storage, away from moisture and light.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Store at -20°C, sealed, away from moisture and light
  • Extended storage in solvent possible at -80°C