Learn More
Medchemexpress LLC M-PEG12-amine | 1977493-48-3 | MFCD00131599 | ≥98.0% | 559.69 g/mol | C25H53NO12 | 100 MG
SDP
Catalog No. 5000203978
Encompass_Preferred
m-PEG12-amine is a methoxy-terminated polyethylene glycol linker containing a 12-unit PEG chain and a terminal primary amine. It serves as a flexible, non-cleavable spacer for conjugation chemistry in linker and ADC/PROTAC synthesis.
  • Methoxy-terminated PEG chain, 12 units.
  • Terminal primary amine suitable for amide or carbamate coupling.
  • Molecular weight approximately 559.7 g/mol; formula C25H53NO12.
  • High purity (≥98%) appropriate for research and synthesis workflows.
  • Supplied in small-scale quantities suitable for laboratory use.
Catalog No. 50-002-03978 Supplier Medchemexpress LLC Supplier No. HY140227100MG
m-PEG12-amine is a methoxy-terminated polyethylene glycol linker containing a 12-unit PEG chain and a terminal primary amine. It serves as a flexible, non-cleavable spacer for conjugation chemistry in linker and ADC/PROTAC synthesis.
  • Methoxy-terminated PEG chain, 12 units.
  • Terminal primary amine suitable for amide or carbamate coupling.
  • Molecular weight approximately 559.7 g/mol; formula C25H53NO12.
  • High purity (≥98%) appropriate for research and synthesis workflows.
  • Supplied in small-scale quantities suitable for laboratory use.