Learn More
Medchemexpress LLC m-PEG12-azide | 2170098-29-8 | 98.6% | C25H51N3O12 | 250 MG
SDP
Catalog No. 5000352722
Encompass_Preferred
m-PEG12-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. This linker helps PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It appears as a colorless to light yellow liquid and is intended for research use only.
  • PEG-based linker for PROTAC synthesis
  • Click chemistry reagent with azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Leverages ubiquitin-proteasome system for target protein degradation
  • Colorless to light yellow liquid appearance
  • Intended for research use only
Catalog No. 50-003-52722 Supplier Medchemexpress LLC Supplier No. 250MGHY140830
m-PEG12-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. This linker helps PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It appears as a colorless to light yellow liquid and is intended for research use only.
  • PEG-based linker for PROTAC synthesis
  • Click chemistry reagent with azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Leverages ubiquitin-proteasome system for target protein degradation
  • Colorless to light yellow liquid appearance
  • Intended for research use only